Anti-proNGF Mouse Monoclonal Antibody (Biotin) [clone: BS312]
Catalog # BSENM-1778-50-B
Gyártó: Biosensis
Specifikációk
- Antibody type:Primary
- Antigen symbol:proNGF
- Clonality:Monoclonal
- Clone:BS312
- Conjugation:Biotin
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:30-41
- Format:IgG
- Form:Lyophilized
- Gene ID:P01138 NGF_Human
- Antigen synonyms:NGF|proNGF|pro-brain nerve growth factor
- Amino acid number:33 to 41
- Antibody fragment:IgG
- Storage buffer:Lyophilised from a solution containing PBS buffer pH 7.4 with 3% trehalose, without preservatives.
- Sequence:HTIPQAHWTKLQ
- Shipping temperature:Ambient
- Immunogen:A synthetic peptide (C-HTIPQAHWTKLQ, aa: 30-41) of human proNGF protein has been used as the immunogen. The sequence is located on the pro-domain of the proNGF full-length protein
- Purification:Antibody was purified from cell culture supernatant by Protein G chromatography, biotinylated and buffer-exchanged into PBS, pH 7.4 buffer
- Size:50 ug
- Pk:50 µG
Specifikációk
Termék információk
Nerve growth factor (NGF) is synthesized as a precursor (proNGF) which may be released and have physiological functions to cause cell death. It binds neurotrophin receptor p75 and sortilin and may also be important for the development of nervous system. proNGF is synthesized in target tissues and glia, transported retrogradely and may be released.
Biosensis' biotinylated proNGF antibody allowing more flexibility in experimental design by using the biotin-avidin/streptavidin detection method. The ability of biotinylated proNGF antibody to detect proNGF has been validated by WB.
Type: Primary
Antigen: proNGF
Clonality: Monoclonal
Clone: BS312
Conjugation: Biotin
Epitope: 30-41
Host: Mouse
Isotype:
Reactivity: Human