Order Entry
Hungary
ContactUsLinkComponent
Anti-RIP Polyclonal Antibody
Anti-RIP Polyclonal Antibody
Catalog # BSBTPB9116
Supplier:  Boster Bio
undefined
Anti-RIP Polyclonal Antibody
Catalog # BSBTPB9116
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    RIP
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Polyclonal antibody for RIP1/RIPK1 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: WB. Reactive species: Human. RIP1/RIPK1 information: Molecular Weight: 75931 MW; Subcellular Localization: Cytoplasm. Cell membrane .

Type: Primary
Antigen: RIP
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human