Disposable samplers, LiquiThief, SteriWare®
Disposable liquid samplers designed for a high number of liquids and low viscosity testing applications.
- No risk of cross- contamination - can be used straight from the bag
- Silicone seal for improved sampling performance
- Assembled and packed in a Class 100 000 cleanroom
Designed for taking fast sterile samples from tanks, kegs, and containers in pharmaceutical, food, chemical, cosmetic, and biotechnology industries.
: FDA, EU 10/2011 and EC 1935/2004 conformity. BSE-/TSE-free. Full batch traceability, ATEX rated.
: Pre-sterilised versions are available.
: Individually bagged in a heat-sealed PE bag.
Frequently Bought Together