Order Entry
Hungary
ContactUsLinkComponent
Gabapentin
Irritant
Health
Gabapentin
Certificates
undefined
Irritant
Health
Gabapentin

Specification Test Results

  • Appearance:
    White to almost white Powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    162 to 168 °C
  • Titration with HClO4:
    ≥97.5 %

Specification Test Results

Frequently Bought Together