Order Entry
Hungary
ContactUsLinkComponent
Methanol with 0,1% (v/v) formic acid, CHROMASOLV™ LC-MS Ultra LabReady™, Solvent Blend tested for UHPLC-MS, Riedel-de Haën™
Health
Flammable
Methanol with 0,1% (v/v) formic acid, CHROMASOLV™ LC-MS Ultra LabReady™, Solvent Blend tested for UHPLC-MS, Riedel-de Haën™
Catalog # 14276-2L
CAS Number:  
Health
Flammable
Methanol with 0,1% (v/v) formic acid, CHROMASOLV™ LC-MS Ultra LabReady™, Solvent Blend tested for UHPLC-MS, Riedel-de Haën™
Catalog # 14276-2L
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2 L
  • Seq:
    5086

Specifications

Specification Test Results

  • Formic acid:
    0.095 - 0.105 Vol.%
  • Non-volatile matter:
    Max. 0.0001 %
  • Calcium (Ca):
    Max. 0.3 ppm
  • Potassium (K):
    Max. 0.3 ppm
  • Magnesium (Mg):
    Max. 0.3 ppm
  • Sodium (Na):
    Max. 1 ppm
  • Transmittance at 254 nm:
    Min. 90 %
  • Transmittance at 230 nm:
    Min. 10 %
  • Suitability for the UHPLC-MS:
    complying
  • UHPLC Gradient drift 254 nm:
    Max. 30 mAU
  • UHPLC Gradient peak 254 nm:
    Max. 4 mAU
  • UHPLC-MS ESI(+) (Reserpine):
    Max. 20 ppb
  • Fluorescence (chinin) at 254 nm:
    Max. 0.7 ppb
  • Fluorescence (chinin) at 365 nm:
    Max. 0.5 ppb

Specification Test Results

About this item

 :
Melting Pt:−46 °C
Flash Pt:11 °C
Storage Temperature:Ambient
EINECS:200-659-6
UN:1230
ADR:3,II
 :GHS08
 :GHS02
 :64-18-6
MDL Number:MFCD00004595