Weighing dishes, aluminium
MARI6604900
- Ext.Ø:100 mm
- Description:Weighing dishes, 80 per pack, 6 packs
- Height:7 mm
- Pk:80
Dishes made of aluminium.
- Flat bottom ensures good standing on the worktop and keeps the samples securely in place
- Manufactured free of grease or oil
- Good chemical and temperature resistance
Suitable for moisture analysis in infra-red dryer.