Order Entry
Hungary
ContactUsLinkComponent
Elemental analysis standards, portland cement type Ⅲ, NIST-1887B
Elemental analysis standards, portland cement type Ⅲ, NIST-1887B
Catalog # PROONIST-1887B
CAS Number:  
Elemental analysis standards, portland cement type Ⅲ, NIST-1887B
Catalog # PROONIST-1887B
CAS Number:  
Specifications
  • Pack type:
    Vial
  • Pk:
    5
  • Description:
    NIST-1887B, Type Ⅲ portland cement
  • Pk Info:
    5× 4 g
Specifications
About this item
These are intended primarily for the evaluation and calibration of methods for analysis of cements and materials of similar matrix.
A unit consists of five vials, each containing at least 4 g of portland cement ground to pass through a 75 µm (No. 200) sieve, and each sealed in a foil pouch.
Certifications: Traceable to NIST and SRM.