Skip to Header
Skip to Main Content
Skip to Footer
Bővített
Bővített
SDS
Tanúsítványok
Gyártói adatok
0
0 Ft
Order Entry
Login
/
Regisztráció
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Hungary
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Kezdőlap
Standards
Organic Standards
Supelco® Drug and metabolite standards, piperine, LoGiCal®
Supelco® Drug and metabolite standards, piperine, LoGiCal®
Supelco® Drug and metabolite standards, piperine, LoGiCal®
Catalog # PROOLGCFOR2522.00
Supplier:
LGC Standards PROMOCHEM
CAS Number:
Expand image
Supelco® Drug and metabolite standards, piperine, LoGiCal®
Catalog # PROOLGCFOR2522.00
Supplier:
LGC Standards PROMOCHEM
CAS Number:
Specifications
Pk:
10 mg
Standards:
Reference standard
Description:
Piperine
Specifications
About this item
Product Details
Product Details