Order Entry
Hungary
ContactUsLinkComponent
Anopheles gambiae recombinant 3-Hydroxykynurenine Transaminase (from E. coli)
Anopheles gambiae recombinant 3-Hydroxykynurenine Transaminase (from E. coli)
Catalog # PRSI90-358
Supplier:  ProSci Inc.
CAS Number:  
Anopheles gambiae recombinant 3-Hydroxykynurenine Transaminase (from E. coli)
Catalog # PRSI90-358
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Enzyme type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Anopheles gambiae
  • Storage conditions:
    Stable for at least 6 months after receipt when stored at –80 °C.
  • Enzyme name:
    3-Hydroxykynurenine Transaminase
  • Enzyme activity:
    Specific activity towards D,L-3HK: 33 umol/min per mg. Primarily catalyses the pyridoxal 5'-phosphate (PLP)-dependent irreversible transamination of 3-hydroxykynurenine (3-HK) to xanthurenic acid (XA).
  • Purity:
    >99% (SDS-PAGE)
  • Formulation:
    Liquid, In 20 mM TRIS-HCl pH 8,0, containing 150 mM NaCl and 40 uM pyridoxal 5'-phosphate (PLP).

Specifications

About this item

The 3-hydroxykynurenine transaminase (3-HKT) gene plays a vital role in the development of malaria parasites by participating in the synthesis of xanthurenic acid, which is involved in the exflagellation of microgametocytes in the midgut of malaria vector species. The 3-HKT enzyme is involved in the tryptophan metabolism of Anophelines. The gene had been studied in the important global malaria vector, Anopheles gambiae.

Fusion-Tag: Tag Free

This recombinant proteins is for research use only.