Order Entry
Hungary
ContactUsLinkComponent
Dechloromonas aromatica (strain RCB) recombinant chlorite dismutase (from E. coli)
Dechloromonas aromatica (strain RCB) recombinant chlorite dismutase (from E. coli)
Catalog # PRSI92-660
Supplier:  ProSci Inc.
CAS Number:  
Dechloromonas aromatica (strain RCB) recombinant chlorite dismutase (from E. coli)
Catalog # PRSI92-660
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Enzyme type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Dechloromonas aromatica (strain RCB)
  • Tag sequence:
    N-6 His tag
  • Storage conditions:
    Lyophilized protein should be stored at –20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4...7 °C for 2 - 7 days. Aliquots of reconstituted samples are stable at –20 °C for 3 months.
  • Enzyme name:
    Chlorite dismutase
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular weight:
    31,3 kDa
  • Sequence:
    Met35-Asp282
  • Formulation:
    Lyophilized from a 0,2 µm filtered solution of PBS, 0,5 mM EDTA, pH 7,4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH₂O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.

Specifications

About this item

Chlorite dismutase (Cld) found in prokaryotic organisms, also known as Chlorite O2-lyase, is a b-type heme containing enzyme that catalyses the reduction of chlorite into chloride plus dioxygen. The subunit of chlorite dismutase consists of a heme free N-terminal and a heme b containing C-terminal ferredoxin-like fold with high structural homology to the dye-decolorizing peroxidases (DyPs). The physiological role of Cld in prokaryote has been shown that some microorganisms can use perchlorate or chlorate as terminal electron acceptors for anaerobic respiration thereby producing chlorite that must be detoxified. This enzyme has gained attention because it can be used in the development of bioremediation processes, biosensors, and controlled dioxygen production.

Fusion-Tag: N-6 His tag

This recombinant protein can be used for biological assays. For research use only.