Order Entry
Hungary
ContactUsLinkComponent
Human recombinant Aldehyde dehydrogenase 3-A1 (from cells)
Human recombinant Aldehyde dehydrogenase 3-A1 (from cells)
Catalog # PRSI92-459
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant Aldehyde dehydrogenase 3-A1 (from cells)
Catalog # PRSI92-459
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Enzyme type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Storage conditions:
    Lyophilized protein should be stored at –20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4...7 °C for 2 - 7 days. Aliquots of reconstituted samples are stable at –20 °C for 3 months.
  • Enzyme name:
    Aldehyde dehydrogenase 3-A1
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular weight:
    51.4 kDa
  • Sequence:
    Met1-His453, C-6 H
  • Formulation:
    Lyophilized from a 0,2 µm filtered solution of PBS,pH 7,4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH₂O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.

Specifications

About this item

Aldehyde dehydrogenase, dimeric NADP-preferring is an enzyme that in humans is encoded by the ALDH3A1 gene, belongs to the aldehyde dehydrogenase family. ALDHs play a major role in the detoxification of alcohol-derived acetaldehyde. They are involved in the metabolism of corticosteroids, biogenic amines, neurotransmitters, and lipid peroxidation. This protein preferentially oxidises aromatic aldehyde substrates. It may play a role in the oxidation of toxic aldehydes.

Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays. For research use only.