Order Entry
Hungary
ContactUsLinkComponent
Human recombinant thymidine kinase 1 (from cells)
Human recombinant thymidine kinase 1 (from cells)
Catalog # PRSI92-369
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant thymidine kinase 1 (from cells)
Catalog # PRSI92-369
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Enzyme type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
  • Enzyme name:
    Thymidine kinase 1
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular weight:
    26.5 kD
  • Sequence:
    Met1-Asn234
  • Formulation:
    Supplied as a 0.2 µm filtered solution of PBS,pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH₂O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Biological assays

Specifications

About this item

Thymidine kinase 1(TK1) belongs to the thymidine kinase family. It is located in the cytoplasm, and phosphorylated on Ser-13 in mitosis during post-translational modification. Two forms of this protein have been identified in animal cells, one in cytosol TK1 and one in mitochondria TK2. Thymidine kinases have a key function in the synthesis of DNA and thereby in cell division, as they are part of the unique reaction chain to introduce deoxythymidine into the DNA. Activity of the cytosolic enzyme is high in proliferating cells and peaks during the S-phase of the cell cycle, while it is very low in resting cells. TK1 acts as a homotetramer, and can transform thymidime to thymidine 5'-phosphate with the help of ATP.

Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays. For research use only.