Order Entry
Hungary
ContactUsLinkComponent
Human recombinant sulphotransferase 4A1 (from E. coli)
Human recombinant sulphotransferase 4A1 (from E. coli)
Catalog # PRSI92-124
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant sulphotransferase 4A1 (from E. coli)
Catalog # PRSI92-124
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Enzyme type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
  • Enzyme name:
    Sulfotransferase 4A1
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular weight:
    33 kD
  • Sequence:
    Met1-Leu284
  • Formulation:
    Supplied as a 0.2 µm filtered solution of 20 mM PB, 150 mM NaCl,pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

Sulfotransferase 4A1 (ST4A1) is a member of the Sulfotransferase 1 family. ST4A1 is highly expressed in the cerebral cortex and frontal lobe, but no expression is detected in the pancreas. ST4A1 is a brain-specific sulfotransferase believed to be involved in the metabolism of neurotransmitters. ST4A1 acts on catecholamines and T4 in a manner that may not involve sulfonation. ST4A1 may have a role in the metabolism of drugs and neurotransmitters in the CNS. In addition, ST4A1 is related to schizophrenia.

This recombinant protein can be used for biological assays. For research use only.