Order Entry
Hungary
ContactUsLinkComponent
Human recombinant CaM Kinase II subunit beta (from E. coli)
Human recombinant CaM Kinase II subunit beta (from E. coli)
Catalog # PRSI92-108
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant CaM Kinase II subunit beta (from E. coli)
Catalog # PRSI92-108
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Enzyme type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
  • Enzyme name:
    CaM Kinase II subunit beta
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular weight:
    57.4 kD
  • Sequence:
    Met1-Gln503
  • Formulation:
    Supplied as a 0.2 µm filtered solution of 10 mM HEPES, 150 mM NaCl, pH 7.5. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

Calcium/Calmodulin-Dependent Protein Kinase Type II Subunit Beta (CAMK2B) is a cytoplasmic protein that belongs to the serine/threonine protein kinase family and the Ca(2+)/calmodulin-dependent protein kinase subfamily. CAMK2B is a calcium/calmodulin-dependent protein kinase that functions autonomously after Ca2+/calmodulin-binding and autophosphorylation. It is involved in dendritic spine and synapse formation, neuronal plasticity and regulation of sarcoplamic reticulum Ca2+ transport in skeletal muscle. In neurons, CAMK2B plays an essential structural role in the reorganisation of the actin cytoskeleton during plasticity by binding and bundling actin filaments in a kinase-independent manner.

Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays. For research use only.