Order Entry
Hungary
ContactUsLinkComponent
 
Human recombinant Lecithin-cholesterol acyltransferase (from cells)
Catalog # PRSI91-786
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant Lecithin-cholesterol acyltransferase (from cells)
Catalog # PRSI91-786
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Enzyme type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Lyophilized protein should be stored at –20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4...7 °C for 2 - 7 days. Aliquots of reconstituted samples are stable at –20 °C for 3 months.
  • Enzyme name:
    Lecithin-cholesterol acyltransferase
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular weight:
    48.1 kD
  • Sequence:
    Phe25-Glu440
  • Formulation:
    Lyophilized from a 0.2 µm filtered solution of 50 mM Acetate Buffer pH-4.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

Lipase family. The gene encoding this protein is expressed mainly in brain, liver and testes,followed by secreting into plasma and cerebral spinal fluid. The esterification of cholesterol is required for cholesterol transport. LCAT is a central enzyme in the extracellular metabolism of plasma lipoproteins. Defects in LCAT are the cause of lecithin-cholesterol acyltransferase deficiency (LCATD) and a cause of fish-eye disease (FED).

Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays. For research use only.