Order Entry
Hungary
ContactUsLinkComponent
Human recombinant trans-2-Enoyl-CoA Reductase (from cells)
Human recombinant trans-2-Enoyl-CoA Reductase (from cells)
Catalog # PRSI91-680
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant trans-2-Enoyl-CoA Reductase (from cells)
Catalog # PRSI91-680
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Enzyme type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Lyophilized protein should be stored at –20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4...7 °C for 2 - 7 days. Aliquots of reconstituted samples are stable at –20 °C for 3 months.
  • Enzyme name:
    Trans-2-Enoyl-CoA Reductase
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular weight:
    35.7 kD
  • Sequence:
    Pro54-Met373
  • Formulation:
    Lyophilized from a 0.2 µm filtered solution of 20 mM PB, 150 mM NaCl,pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

Trans-2-Enoyl-CoA Reductase Mitochondrial (MECR) belongs to the zinc-containing alcohol dehydrogenase family. MECR localises to the mitochondrion. It is highly expressed in skeletal and heart muscle and expressed at lower levels in the placenta, liver, kidney and pancreas, with weakly or no expression in the lung. MECR exists as a homodimer, which catalyses the reduction of trans-2-enoyl-CoA to acyl-CoA with chain length from C6 to C16 in an NADPH-dependent manner with preference to medium chain length substrate. MECR may take part in the mitochondrial synthesis of fatty acids.

Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays. For research use only.