Human recombinant fumarylacetoacetase (from cells)
Catalog # PRSI91-668
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Enzyme type:Recombinant
- Source:Cells
- Species:Human
- Tag sequence:C-6 His tag
- Storage conditions:Lyophilized protein should be stored at –20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4...7 °C for 2 - 7 days. Aliquots of reconstituted samples are stable at –20 °C for 3 months.
- Enzyme name:Fumarylacetoacetase
- Purity:>95% as determined by reducing SDS-PAGE
- Molecular weight:47.4 kD
- Sequence:Ser2-Ser419
- Formulation:Lyophilized from a 0.2 µm filtered solution of PBS. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
Specifications
About this item
Fumarylacetoacetase belongs to the FAH family. Fumarylacetoacetase is primary expressed in liver and kidney. It exists as a homodimer and catalyses the hydrolysis of 4-fumarylacetoacetate into fumarate and acetoacetate. Defects in Fumarylacetoacetase cause tyrosinemia type 1, which is congenital metabolism defect characterised by elevated levels of tyrosine in the blood and urine, and hepatorenal manifestations. Typical features include renal tubular injury, self-mutilation, hepatic necrosis, episodic weakness, and seizures.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.