Human recombinant Pancreatic Lipase-Related 1 (from cells)
PRSI91-568
:
- Pk:50 µG
- Enzyme type:Recombinant
- Source:Cells
- Species:Human
- Tag sequence:C-6 His tag
- Storage conditions:Lyophilized protein should be stored at –20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4...7 °C for 2 - 7 days. Aliquots of reconstituted samples are stable at –20 °C for 3 months.
- Enzyme name:Pancreatic Lipase-Related 1
- Purity:>95% as determined by reducing SDS-PAGE
- Molecular weight:50.99 kD
- Sequence:Lys18-Cys467
- Formulation:Lyophilized from a 0.2 µm filtered solution of 20 mM PB, 150 mM NaCl, pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
Pancreatic Lipase-Related Protein 1 (PNLIPRP1) belongs to the Lipase family within the AB hydrolase superfamily. PNLIPRP1 is a secreted protein and contains one PLAT domain. PNLIPRP1 is involved in lipid metabolic process, acting as a negative regulator of pancreatic lipase activity by competing with pancreatic lipase for colipase occupancy. PNLIPRP1 may play a role in inhibiting dietary triglyceride digestion, but it lacks detectable lipase activity towards triglycerides, diglycerides, phosphatidylcholine, galactolipids or cholesterol esters.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.