Order Entry
Hungary
ContactUsLinkComponent
Human recombinant beta-Galactosidase (from Cells)
Human recombinant beta-Galactosidase (from Cells)
Catalog # PRSI91-428
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant beta-Galactosidase (from Cells)
Catalog # PRSI91-428
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Enzyme type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
  • Enzyme name:
    beta-Galactosidase
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular weight:
    74.63 kD
  • Sequence:
    Leu24-Val677
  • Formulation:
    Supplied as a 0.2 µm filtered solution of 20 mM TrisHCl, 150 mM NaCl, pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    For Biological Assays

Specifications

About this item

beta Galactosidase is a lysosomal beta Galactosidase that hydrolyses the terminal beta Galactose from Ganglioside and Keratan sulfate. In lysosome, the mature beta Galactosidase protein associates with Cathepsin A and Neuraminidase 1 to form the lysosomal multienzyme complex . An alternative splicing at the RNA level of beta Galactosidase results a catalytically inactive beta Galactosidase that plays an important role in vascular development. Defects of beta -galactosidase (GLB1) are the cause of diseases like GM1-gangliosidosis which is a lysosomal storage disease and Morquio Syndrome B that cause patients to have abnormal elastic fibers. More than 100 mutations have been identified for beta Galactosidase, which result in different residual activities of the mutant enzymes and a spectrum of symptoms in the two related diseases.

Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays. For research use only.