Order Entry
Hungary
ContactUsLinkComponent
Human recombinant QAPRTase (from E. coli)
Human recombinant QAPRTase (from E. coli)
Catalog # PRSI91-237
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant QAPRTase (from E. coli)
Catalog # PRSI91-237
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Enzyme type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Tag sequence:
    N-6 His tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
  • Enzyme name:
    QAPRTase
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular weight:
    33 kD
  • Sequence:
    Met1-His297
  • Formulation:
    Supplied as a 0.2 µm filtered solution of 20 mM TrisHCl, 150 mM NaCl, pH 8.0 . Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.

Specifications

About this item

Nicotinate-Nucleotide Pyrophosphorylase (QPRT) belongs to the nadC/modD family. QPRT plays an improtant role in catabolism of quinolinate which acts as a potent endogenous exitotoxin to neurons. In addition, QPRT serves as an an intermediate in the Tryptophan-Nicotinamide Adenine Dinucleotide pathway. QPRT participates in some pathways including Cofactor biosynthesis, NAD(+) biosynthesis and the Nicotinate D-Ribonucleotide from Quinolinate. In addition, QPRT is involved in the catabolism of Quinolinic Acid (QA). The activity toward QA is slightly repressed by phosphoribosylpyrophosphate (PRPP) in both a competitive and a non-competitive manner.

Fusion-Tag: N-6 His tag

This recombinant protein can be used for biological assays. For research use only.