Order Entry
Hungary
ContactUsLinkComponent
Human recombinant PMKASE (from E. coli)
Human recombinant PMKASE (from E. coli)
Catalog # PRSI91-228
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant PMKASE (from E. coli)
Catalog # PRSI91-228
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Enzyme type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Tag sequence:
    N-6 His tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
  • Enzyme name:
    PMKASE
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular weight:
    24.2 kD
  • Sequence:
    Met1-Leu192
  • Formulation:
    Supplied as a 0.2 µm filtered solution of 20 mM TrisHCl, 100 mM NaCl, 1 mM DTT, 10% Glycerol, pH 7.5. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

Phosphomevalonate kinase (PMVK) is a cytosolic enzyme. PMVK can be highly expressed in the heart,skeletal muscle, liver, pancreas, and kidney; it is expressed at lower levels in the brain, lung, and placenta. Induced by sterol, PMVK takes part in isopentenyl diphosphate biosynthesis through the mevalonate pathway. PMVK catalyses the conversion of mevalonate 5-phosphate into mevalonate 5-diphosphate in the fifth reaction of the cholesterol biosynthetic pathway.

Fusion-Tag: N-6 His tag

This recombinant protein can be used for biological assays. For research use only.