Order Entry
Hungary
ContactUsLinkComponent
Human recombinant Aldolase A (from E. coli)
Human recombinant Aldolase A (from E. coli)
Catalog # PRSI91-182
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant Aldolase A (from E. coli)
Catalog # PRSI91-182
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Enzyme type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
  • Enzyme name:
    Aldolase A
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular weight:
    40.48 kD
  • Sequence:
    Pro2-Tyr364
  • Formulation:
    Supplied as a 0.2 µm filtered solution of 20 mM TrisHCl, 100 mM NaCl, 20% Glycerol, pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    For Biological assays

Specifications

About this item

Fructose Bisphosphate Aldolase A (ALDOA) belongs to the class I fructose-bisphosphate aldolase family. ALDOA is a glycolytic enzyme that catalyses the reversible conversion of fructose-1,6-bisphosphate to glyceraldehyde 3-phosphate and dihydroxyacetone phosphate. In vertebrates, three forms of this ubiquitous glycolytic enzyme are found, Aldolase A in muscle, Aldolase B in liver and aldolase C in brain. Aldolase A Interacts with SNX9 and WAS. Aldolase A deficiency has been associated with myopathy and hemolytic anemia. In addition, Aldolase A plays an important role in glycolysis and gluconeogenesis; it may also act as a scaffolding protein.

Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays. For research use only.