Human recombinant N-Glycosylase (from E. coli)
Catalog # PRSI91-111
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Enzyme type:Recombinant
- Source:E. coli
- Species:Human
- Tag sequence:N-GST tag
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
- Enzyme name:N-Glycosylase
- Purity:>95% as determined by reducing SDS-PAGE
- Molecular weight:65.07 kD
- Sequence:Met1-Gly345
- Formulation:Supplied as a 0.2 µm filtered solution of 20 mM TrisHCl, 100 mM NaCl, 1 mM DTT, 1 mM EDTA, 50% Glycerol, pH 7.8. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
Specifications
About this item
Human N-Glycosylase/DNA Lyase is a DNA repair enzyme, which incises DNA at 8-oxoG residues, and excises 7,8-dihydro-8-oxoguanine and 2,6-diamino-4-hydroxy-5-N-methylformamidopyrimidine (FAPY) from damage DNA. DGG1 has a beta -lyase activity that nicks DNA 3’ to the lesion.
Fusion-Tag: N-GST tag
This recombinant protein can be used for biological assays. For research use only.