Jugs, PP, SteriWare®
Made from PP (FDA, EC 1935/2004 and EU 10/2011 conforming).
- Moulded and packed in a cleanroom
- BSE/TSE free
- Full batch traceability
- Graduation marks at every 10 ml
These range of class leading jugs are available pre-sterilised (gamma irradiated). The ergonomic handle is rounded to make it comfortable to hold and use.
: Individually bagged.
Frequently Bought Together