Order Entry
Hungary
Orders LinkContactUsLinkComponent
Bleached Shellac, Reference Standards
Bleached Shellac, Reference Standards
Supplier:   USP
Bleached Shellac, Reference Standards
Supplier:  USP
Certificates
2 Options Available Below
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
Specifications for Product Family
Specifications
Specifications
  • Catalog No:
  • 1600314.
  • 1600303.
  • Description:
  • Regular Bleached Shellac
  • Refined Bleached Shellac
  • CAS No.:
  • 9000-59-3
  • 9000-59-3
Specifications
Product Family Options
Product Information
  • Standards
    Pack type
    Pk
    Availability
    Price
  • 1600314.
    Regular Bleached Shellac
    Regular Bleached Shellac
    USP reference standard
    Vial
    100 mg
    Log in for availability
    Log in for availability
    1 * 100 mg
    1 * 100 mg - 85 500 Ft
    85 500 Ft
    1 * 100 mg
       85 500 Ft
    Specifications:
    More
    Specifications
    Cat. No.
    1600314.
    Description
    Regular Bleached Shellac
    CAS No.
    9000-59-3
  • 1600303.
    Refined Bleached Shellac
    Refined Bleached Shellac
    USP reference standard
    Vial
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 156 000 Ft
    156 000 Ft
    1 * 1 g
       156 000 Ft
    Specifications:
    More
    Specifications
    Cat. No.
    1600303.
    Description
    Refined Bleached Shellac
    CAS No.
    9000-59-3