Order Entry
Hungary
Orders LinkContactUsLinkComponent
 

Bicalutamide, Reference Standards

Supplier:   USP
Bicalutamide, Reference Standards
Supplier:  USP
Certificates

3 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

 : US Pharmacopea Reference Standard

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1071202.
  • 1071213.
  • 1071224.
  • Description:
  • Bicalutamide
  • Bicalutamide Related Compound A
  • Bicalutamide Related Compound B
  • Synonym(s):
  • N-[4-cyano-3-(trifluoromethyl)phenyl]-3-[(4-fluoro-phenyl)sulFood Ingredientsnyl]-2-hydroxy-2-methyl-propanamide
  • (RS)-N-(4-Cyano-3-(trifluoromethyl)phenyl)-3-(3-fluorophenylsulfonyl)-2-hydroxy-2-methylpropanamide
  • CAS No.:
  • 90357-06-5
  • 1166228-30-3
  • Formula:
  • C₁₈H₁₄F₄N₂O₄S
  • C₁₈H₁₄F₄N₂O₃S
  • C₁₈H₁₄F₄N₂O₄S
  • UN No.:
  • 3077
  • 3077

Specifications

Product Family Options

Product Information

  • Standards
    Pack type
    Pk
    Availability
    Price
  • 1071202.
    Bicalutamide
    Bicalutamide
    USP reference standard
    Vial
    200 mg
    Log in for availability
    Log in for availability
    1 * 200 mg
    1 * 200 mg - 152 000 Ft
    152 000 Ft
    1 * 200 mg
       152 000 Ft

    Specifications:
    More
    Specifications
    Cat. No.
    1071202.
    Description
    Bicalutamide
    Synonym(s)
    CAS No.
    90357-06-5
    Formula
    C₁₈H₁₄F₄N₂O₄S
    UN No.
    3077
  • 1071213.
    Bicalutamide Related Compound A
    Bicalutamide Related Compound A
    USP reference standard
    Vial
    25 mg
    Log in for availability
    Log in for availability
    1 * 25 mg
    1 * 25 mg - 449 000 Ft
    449 000 Ft
    1 * 25 mg
       449 000 Ft

    Specifications:
    More
    Specifications
    Cat. No.
    1071213.
    Description
    Bicalutamide Related Compound A
    Synonym(s)
    N-[4-cyano-3-(trifluoromethyl)phenyl]-3-[(4-fluoro-phenyl)sulFood Ingredientsnyl]-2-hydroxy-2-methyl-propanamide
    CAS No.
    Formula
    C₁₈H₁₄F₄N₂O₃S
    UN No.
  • 1071224.
    Bicalutamide Related Compound B
    Bicalutamide Related Compound B
    USP reference standard
    Vial
    10 mg
    Log in for availability
    Log in for availability
    1 * 10 mg
    1 * 10 mg - 507 000 Ft
    507 000 Ft
    1 * 10 mg
       507 000 Ft

    Specifications:
    More
    Specifications
    Cat. No.
    1071224.
    Description
    Bicalutamide Related Compound B
    Synonym(s)
    (RS)-N-(4-Cyano-3-(trifluoromethyl)phenyl)-3-(3-fluorophenylsulfonyl)-2-hydroxy-2-methylpropanamide
    CAS No.
    1166228-30-3
    Formula
    C₁₈H₁₄F₄N₂O₄S
    UN No.
    3077
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.