Order Entry
Hungary
ContactUsLinkComponent
 
Silicone antifoam, liquid (ORG), Technical Grade
  APPC211628.1210
 :  
Silicone antifoam, liquid (ORG), Technical Grade
  APPC211628.1210
 :  

 

  • Pk:
    500 mL
  • Seq:
    2725

 

 

  • Form:
    Liquid
  • Colour:
    Off-white
  • Solubility:
    Immiscible with water

 

 

 :
 :0.97 kg/l
 :Ambient