Impurity and API standard, (2RS)-2-phenyl-2,3-dihydro-4H-1-benzopyran-4-one
Catalog # PROOMM0405.09
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pk:100 mg
- Standards:Reference Standards
- CAS No.:487-26-3
- Description:(2RS)-2-Phenyl-2,3-dihydro-4H-1-benzopyran-4-one
- Formula:C₁₅H₁₂O₂
- Description:(2RS)-2-Phenyl-2,3-dihydro-4H-1-benzopyran-4-one