Order Entry
Hungary
ContactUsLinkComponent
 
Impurity and API standard, droperidol
  PROOMM0138.00
 :  
Impurity and API standard, droperidol
  PROOMM0138.00
 :  

 

  • Pk:
    250 mg
  • Standards:
    Reference standards
  • CAS No.:
    548-73-2
  • Description:
    Droperidol
  • Formula:
    C₂₂H₂₂FN₃O₂
  • Description:
    Droperidol