Order Entry
Hungary
ContactUsLinkComponent
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) ≥99.0% (dried basis), BAKER ANALYZED®, Biochemical Reagent for liquid chromatography, J.T.Baker®
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) ≥99.0% (dried basis), BAKER ANALYZED®, Biochemical Reagent for liquid chromatography, J.T.Baker®
Catalog # 4099-07
Supplier:  Avantor
CAS Number:  
undefined
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) ≥99.0% (dried basis), BAKER ANALYZED®, Biochemical Reagent for liquid chromatography, J.T.Baker®
Catalog # 4099-07
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    PE pail, screw top
  • Pk:
    12 kg
  • Seq:
    4187

Specifications

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    ≥99.0%
  • Appearance:
    Passes Test
  • Assay (C₄H₁₁NO₃) (dried basis):
    ≥99.0 %
  • Absorbance of a 40% Solution - 280 nm:
    ≤0.200
  • Absorbance of a 40% Solution - 400 nm:
    ≤0.020
  • Insoluble Matter:
    ≤0.005 %
  • Water (H₂O)(by Karl Fischer titrn):
    ≤2 %
  • Trace Impurities - ACS - Heavy Metals (as Pb):
    ≤5 ppm
  • Trace Impurities - Iron (Fe):
    ≤5 ppm

Specification Test Results

About this item

 :
 :121.14 g/mol
Merck Index:13,09842
Boiling Pt:357 °C (1013 hPa)
Melting Pt:169…172 °C
Density:1.328 g/cm³ (20 °C)
 :Management of Change (MOC) category = N
Storage Temperature:Ambient
EINECS:201-064-4
CAS Number:77-86-1
Formula:NH₂C(CH₂OH)₃
MDL Number:MFCD00004679