Anti-BMP15 Rabbit Polyclonal Antibody (Cy5®)
Catalog # BOSSBS-4720R-CY5
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Bone Morphogenetic Protein 15
- Antigen symbol:BMP15
- Clonality:Polyclonal
- Conjugation:Cy5®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:2661
- Antigen synonyms:ODG2|BMP-15|GDF9B|POF4|Bone morphogenetic protein 15
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:321-371/454
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Required for ovarian folliculogenesis. Promotes primordial follicle development. Stimulates granulosa cell proliferation. Promotes cell transition from G0/G1 to S and G2/M phases, through an increase of CCND1 and CCNE1 expression, and RB1 phosphorylation. It regulates STAR expression and cAMP-dependent progesterone release in granulosa and thecal cells. Attenuates the suppressive effects of activin A on STAR expression and progesterone production by increasing the expression of inhibin B. It suppresses FST and FSTL3 production in granulosa-lutein cells.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: BMP15
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat