Order Entry
Hungary
ContactUsLinkComponent
Anti-NANOG Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Anti-NANOG Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Catalog # BOSSBS-0829R-A647
Supplier:  Bioss
Anti-NANOG Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Catalog # BOSSBS-0829R-A647
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Homeobox protein NANOG
  • Antigen symbol:
    NANOG
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 647
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    71950
  • Antigen synonyms:
    ENK|ecat4|2410002E02Rik
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Immunogen:
    90-140/305
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

Transcription regulator involved in inner cell mass and embryonic stem (ES) cells proliferation and self-renewal. Imposes pluripotency on ES cells and prevents their differentiation towards extraembryonic endoderm and trophectoderm lineages. Blocks bone morphogenetic protein-induced mesoderm differentiation of ES cells by physically interacting with SMAD1 and interfering with the recruitment of coactivators to the active SMAD transcriptional complexes. Acts as a transcriptional activator or repressor. Binds optimally to the DNA consensus sequence 5'-TAAT[GT][GT]-3' or 5'-[CG][GA][CG]C[GC]ATTAN[GC]-3'. Able to autorepress its expression in differentiating (ES) cells: binds to its own promoter following interaction with ZNF281/ZFP281, leading to recruitment of the NuRD complex and subsequent repression of expression. When overexpressed, promotes cells to enter into S phase and proliferation.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: NANOG
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat