Anti-STARD5 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Catalog # BOSSBS-3919R-A350
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:StAR-related lipid transfer (START) Domain Containing 5
- Antigen symbol:STARD5
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 350
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:80765
- Antigen synonyms:StAR related lipid transfer protein 5|START domain containing 5|MGC10327.|StARD5|START domain containing protein 5
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
STARD5, or START domain containing 5, belongs to a family containing a 200- to 210-amino acid STAR-related lipid transfer (START) domain. STARD5 may be involved in the intracellular transport of sterols or other lipids and may bind cholesterol or other sterols.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: STARD5
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat