Order Entry
Hungary
ContactUsLinkComponent
 
Gas sampling bags, tedlar®,
Catalog # VARI99636003
Supplier:  VARIAN
Gas sampling bags, tedlar®,
Catalog # VARI99636003
Supplier:  VARIAN

Specifications

  • Capacity:
    4 L
  • Description:
    Gas sampling bag
  • Seq:
    02
  • Pk:
    1
  • W×L:
    310×310 mm

Specifications

About this item

Gas sampling bags in Tedlar.