Order Entry
Hungary
ContactUsLinkComponent
 
Human FGF-19 ELISA kit
Catalog # BIRBORB54823-96
Supplier:  Biorbyt
CAS Number:  
Human FGF-19 ELISA kit
Catalog # BIRBORB54823-96
Supplier:  Biorbyt
CAS Number:  

Specifications

  • Pk:
    96
  • Primary antibody reactivity:
    Human
  • Description:
    Human FGF-19 ELISA Kit
  • Detection range:
    31,2 - 2000 pg/ml

Specifications

About this item

Human FGF-19 ELISA kit for the quantitative determination of human fibroblast growth factor 19 (FGF-19).

FGF-19 is a member of a subfamily of FGFs that includes FGF-21 and FGF-23, each member functions as an important regular of nutrient metabolism. The primary source of endocrine FGF-19 is the ileum, bile acids release into the intestine after a meal to induce expression of FGF-19. Circulating FGF-19 plays an important role in maintaining proper bile acid homeostasis. Several pharmacologic studies in hyperglycaemic, obese animal models have shown that FGF-19 can improve metabolic rate and lower serum glucose and hepatic triglyceride and cholesterol levels. Like insulin, FGF-19 functions as postprandial hormone to govern hepatic protein synthesis, glycogen synthesis and gluconeogenesis, but does not stimulate lipogenesis.