Order Entry
Hungary
ContactUsLinkComponent
Human CXCL13 ELISA kit
Human CXCL13 ELISA kit
Catalog # BIRBORB106615-96
Gyártó:  Biorbyt
CAS szám:  
undefined
Human CXCL13 ELISA kit
Catalog # BIRBORB106615-96
Gyártó:  Biorbyt
CAS szám:  

Specifikációk

  • Pk:
    96
  • Primary antibody reactivity:
    Human
  • Description:
    Human CXCL13 ELISA Kit
  • Detection range:
    15,6 - 1000 pg/ml
  • Sensitivity:
    <1 pg/ml

Specifikációk

Termék információk

Human CXCL13 ELISA Kit for the quantitative determination of human Chemokine (C-X-C motif) 13 (CXCL13).

CXCL13, also known as B lymphocyte chemoattractant (BLC), is a protein that in humans is encoded by the CXCL13 gene. CXCL13 is a small cytokine belonging to the CXC chemokine family, and it was located on sequence maps to 4q21, near the genes encoding most CXC chemokines. In T-lymphocytes, CXCL13 expression is thought to reflect a germinal centre origin of the T-cell. Hence, expression of CXCL13 in T-cell lymphomas, such as Angioimmunoblastic T-cell Lymphoma, is thought to reflect a germinal center origin of the neoplastic T-cells.