Order Entry
Hungary
ContactUsLinkComponent
Rat CCL4 ELISA kit
Rat CCL4 ELISA kit
Catalog # BIRBORB106608-96
Supplier:  Biorbyt
CAS Number:  
undefined
Rat CCL4 ELISA kit
Catalog # BIRBORB106608-96
Supplier:  Biorbyt
CAS Number:  

Specifications

  • Pk:
    96
  • Primary antibody reactivity:
    Rat
  • Description:
    Rat CCL4 ELISA kit, 96-well
  • Detection range:
    15,6 - 1000 pg/ml
  • Sensitivity:
    <10 pg/ml

Specifications

About this item

Chemokine (C-C motif) ligand 4, also known as CCL4, is a protein which in mouse is encoded by the CCL4 gene. It is a CC chemokine with specificity for CCR5 receptors.

It is a chemoattractant for natural killer cells, monocytes and a variety of other immune cells. CCL4 is a major HIV-suppressive factor produced by CD8+ T cells. Performing-low memory CD8+ T cells that normally synthesise MIP-1-beta.

By analysis of somatic cell hybrids and by in situ hybridization, the CCL4 gene was assigned to 17q21-q23. Based on functional analysis of MIP-1, one or more additional chemokine receptors that transduce MIP-1 signals must also exist.