Human calbindin 2 (from E. coli)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Source:E. coli
- Species:Human
- Storage conditions:CALB2 although stable 4 °C for weeks, should be stored desiccated below −18 °C. For long term storage it is recommended to add carrier protein (0.1% HSA or BSA).Please prevent freeze-thaw cycles.
- Protein synonyms:Calretinin
- Protein/peptide name:Calbindin 2
- Purity:Greater than 95% as determined by SDS-PAGE.
- Sequence:MGSSHHHHHH SSGLVPRGSH MAGPQQQPPY LHLAELTASQ FLEIWKHFDA DGNGYIEGKE LENFFQELEK ARKGSGMMSK SDNFGEKMKEFMQKYDKNSD GKIEMAELAQ ILPTEENFLL CFRQHVGSST EFMEAWRKYD TDRSGYIEAN ELKGFLSDLL KKANRPYDEP KLQEYTQTIL RMFDLNGDGK LGLSEMSRLL PVQENFLLKF QGMKLTSEEF NAIFTFYDKD RSGYIDEHEL DALLKDLYEK NKKEMNIQQL TNYRKSVMSLAEAGKLYRKD LEIVLCSEPP M.
- Formulation:The CALB2 1mg/ml protein solution contains 20 mM Tris-HCl buffer pH-8 10% glycerol.
- Tested applications:SDS-PAGE , HPLC , FuncS
Specifications
Frequently Bought Together