Human MIF (from E. coli)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Source:E. coli
- Species:Human
- Storage conditions:Liquid MIF although stable 4 °C for week, should be stored desiccated below −18 °C. Please prevent freeze/thaw cycles.
- Protein synonyms:acrophage migration inhibitory factor (glycosylation-inhibiting factor)
- Protein/peptide name:MIF
- Purity:Greater than 95.0% as determined by(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
- Sequence:MRGSHHHHHH GMASMTGGQQ MGRDLYDDDD KDRWGSMPMF IVNTNV PRA SVPDGFL SE LTQQLAQA T GKPPQYIAV HVVPDQLMAF GGSSEPCAL C LHSIGKIG GA QNRSYSK LLC GLLAER LRIS PDRVY INYYD MNAA NVGWNN STF A.
- Formulation:1mg/ml solution containing PBS pH-7.4.
- Tested applications:SDS-PAGE
Specifications
Frequently Bought Together