Order Entry
Hungary
ContactUsLinkComponent
SARS nucleocaspid (fromE. coli)
SARS nucleocaspid (fromE. coli)
Supplier:  Biorbyt
Certificates
SARS nucleocaspid (fromE. coli)
Supplier:  Biorbyt

Specifications

  • Source:
    E. coli
  • Storage conditions:
    Aliquot and store at −20 °C or −80 °C. Avoid repeated freeze/thaw cycles.
  • Protein/peptide name:
    SARS Nucleocaspid
  • Purity:
    Greater than 95% as determined by SDS-PAGE. Three-step procedure using affinity Ni-NTA chromatography and size exclusion chromatography before and after refolding.
  • Solubility:
    Add 0.2 uml of 0.1M Acetate buffer pH4 and let the lyophilized pellet dissolve completely. For conversion into higher pH value, we recommend intensive dilution by relevant buffer to concentration of 10ug/ml. In higher concentrations the solubility of this
  • Formulation:
    Sterile filtered and lyophilized from 0.5 mg/ml in 0.05 Acetate buffer pH4.
  • Tested applications:
    SDS-PAGE

Specifications

Related Information

Frequently Bought Together