Order Entry
Hungary
ContactUsLinkComponent
Human haptoglobin (from E. coli)
Human haptoglobin (from E. coli)
Supplier:  Biorbyt
Certificates
Human haptoglobin (from E. coli)
Supplier:  Biorbyt

Specifications

  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    Lyophilized Haptoglobin although stable at room temperature for weeks, should be stored desiccated below −18 °C. Upon reconstitution Haptoglobin should be stored at 4 °C between 2-7 days and for future use below −18 °C. For long term storage it is recommended to add carrier protein (0. 1% HSA or BSA). Please prevent freeze/thaw cycles.
  • Protein synonyms:
    Haptoglobin alpha chain
  • Protein/peptide name:
    Haptoglobin
  • Purity:
    Greater than 90.0% as determined by SDS-PAGE and HPLC.
  • Solubility:
    It is recommended to reconstitute the lyophilized Haptoglobin in sterile H2O not less than 100ug/ml, which can then be further diluted to other aqueous solutions.
  • Formulation:
    Each mg was lyophilized with 1xPBS, 0.1% SDS and 1mM DTT.
  • Tested applications:
    SDS-PAGE

Specifications

Related Information

Frequently Bought Together