NKp46 (from E. coli)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Source:E. coli
- Storage conditions:Store at 4 °C if entire vial will be used within 2-4 weeks. Store frozen at −20 °C for longer periods of time. For long term storage it is recommended to add carrier protein (0. 1% HSA or BSA). Avoid multiple freeze/thaw cycles.
- Protein/peptide name:NKp46
- Purity:Greater than 95.0% as determined by RP-HPLC and analysis by SDS-PAGE
- Sequence:MQQQTLPKPF IWAEPHFMVP KEKQVTICCQ GNYGAVEYQL HFEGSLFAVD RPKPPERINKVKFYIPDMNS RMAGQYSCIY RVGELWSEPS NLLDLVVTEM YDTPTLSVHP GPEVISGEEV TFYCRLDTAT SMFLLLKEGR SSHVQRGYGK VQAEFPLGPV TTAHRGTYRX FGSYNNHAWSFPSEPVKLLV TGDIENTSLA PEDPTFSADT WGTYLLTTET GLQKDHALWD HTAQN.
- Formulation:The protein contains phosphate buffered saline (pH7.4) 1mM EDTA.
- Tested applications:SDS-PAGE
Specifications
Frequently Bought Together