Human recombinant BOLA1 (from E. coli)
BIRBORB81250-5
: Biorbyt
:
- Pk:5 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:For long term storage store at −20 °C. Avoid freeze/thaw cycles.
- Biological activity:BOLA1 is active in DNA binding assays.
- Protein synonyms:BolA-like
- Protein/peptide name:BOLA1
- Purity:Greater than 95.0% as determined by SDS-PAGE.
- Molecular weight:14kDa
- Sequence:MGSSHHHHHH SSGLVPRGSH MGSQGSAGSG AIGPVEAAIR TKLEEALSPE VLELRNESGG HAVPPGSETH FRVAVVSSRF EGLSPLQRHR LVHAALAEEL GGPVHALAIQ ARTPAQWREN SQLDTSPPCL GGNKKTLGTP
- Formulation:BOLA1 protein solution (1mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.2M NaCl, 20% glycerol and 1mM DTT.
- Tested applications:SDS-PAGE