Human kinase AKT alpha inactive enzyme (from Insect cells (Sf9))
Supplier: Biorbyt
Certificates
About this item
Specifications
- Source:Insect cells (Sf9)
- Species:Human
- Storage conditions:Aliquot and store at −20 °C or −80 °C. Avoid repeated freeze/thaw cycles.
- Biological activity:No protease activity (Twinning test). The Specific activity is 235 U/mg.
- Protein synonyms:AKT 1
- Protein/peptide name:Kinase AKT alpha Inactive Enzyme
- Purity:Greater than 90% as determined by SDS-PAGE.
- Formulation:PKAkt1 1.9mg/ml, in 50mM Tris-HCl, 100mM NaCl, 1mM DTT, 25mM beta glycerophosphate, 50% glycerol, pH 8.5.
- Tested applications:SDS-PAGE
Specifications
Frequently Bought Together