Order Entry
Hungary
ContactUsLinkComponent
Human recombinant Galectin 3 (from E. coli)
Human recombinant Galectin 3 (from E. coli)
Catalog # BIRBORB80331-5
Supplier:  Biorbyt
CAS Number:  
Human recombinant Galectin 3 (from E. coli)
Catalog # BIRBORB80331-5
Supplier:  Biorbyt
CAS Number:  

Specifications

  • Pk:
    5 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    Store at 4 °C if entire vial will be used within 2-4 weeks. Store frozen at −20 °C for longer periods of time. Please avoid freeze thaw cycles.
  • Protein synonyms:
    CBP 35
  • Protein/peptide name:
    Galectin 3
  • Formulation:
    Galectin-3 is supplied in 50mM Tris-Acetate, pH-7.5, 1mM EDTA and 20% Glycerol.
  • Tested applications:
    SDS-PAGE , HPLC , FuncS

Specifications

About this item