Human PDGF B (from E. coli)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Source:E. coli
- Species:Human
- Storage conditions:Store at 4 °C if entire vial will be used within 2-4 weeks. Store frozen at −20 °C for longer periods of time. Please avoid freeze thaw cycles.
- Protein/peptide name:PDGF B
- Purity:Greater than 95.0% as determined by SDS-PAGE.
- Formulation:Platelet-Derived Growth Factor-B is supplied in 0.5M Urea, 20 mM Tris HCl pH8 and 50% glycerol.
- Tested applications:SDS-PAGE
Specifications
Frequently Bought Together