Human CSNK2A1, CK alpha (from E. coli)
: Biorbyt
- Source:E. coli
- Species:Human
- Storage conditions:Aliquot and store at −20 °C or −80 °C. Avoid repeated freeze/thaw cycles.
- Protein synonyms:EC 27111
- Protein/peptide name:CSNK2A1, CK alpha
- Purity:Greater than 99% as determined by SDS-PAGE.
- Formulation:CSNK2A1 is supplied in 25mM Tris-HCl, 500mM NaCl, 1mM DTT, 500um PMSF, 50% glycerol, pH 8.5.
- Tested applications:SDS-PAGE , HPLC , FuncS
Frequently Bought Together