Order Entry
Hungary
ContactUsLinkComponent
Human recombinant Monocyte Chemotactic Protein 1 (from cells)
Human recombinant Monocyte Chemotactic Protein 1 (from cells)
Catalog # BIRBORB81871-20
Supplier:  Biorbyt
CAS Number:  
Human recombinant Monocyte Chemotactic Protein 1 (from cells)
Catalog # BIRBORB81871-20
Supplier:  Biorbyt
CAS Number:  

Specifications

  • Pk:
    20 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Protein synonyms:
    MCAF|MCP-1
  • Protein/peptide name:
    Monocyte Chemotactic Protein 1
  • Purity:
    Purified.
  • Formulation:
    Lyophilised.

Specifications

About this item