Human recombinant HPG1 (from E. coli)
Gyártó: Biorbyt
Tanúsítványok
Termék információk
Specifikációk
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:Store lyophilized protein at −20 °C. Aliquot the product after reconstitution to avoid repeated freezing/thawing cycles. Reconstituted protein can be stored at 4 °C for limited period of time.The lyophilized protein remains stable until the expiry date when stored at −20 °C.
- Protein synonyms:Prostate-Specific Gene-1
- Protein/peptide name:HPG1
- Purity:Greater than 95% as determined by SDS-PAGE. Ni-NTA affinity chromatography.
- Solubility:Add 0.2 uml of dH2O and let the lyophilized pellet dissolve completely. For conversion into higher pH value, we recommend intensive dilution by relevant buffer to concentration of 10ug/ml. In higher concentrations the solubility of this antigen is limited
- Formulation:Sterile filtered and lyophilized from 0.5 mg/ml in 0.05M MES buffer pH 6.5.
- Tested applications:SDS-PAGE
Specifikációk
Frequently Bought Together