Human recombinant PKC-a (from insect cells (Sf9))
Supplier: Biorbyt
Certificates
About this item
Specifications
- Protein/peptide type:Recombinant
- Source:Insect cells (Sf9)
- Species:Human
- Storage conditions:Aliquot and store at −20 °C or −80 °C. Avoid repeated freeze/thaw cycles.
- Protein synonyms:Protein kinase alpha type
- Protein/peptide name:PKC-a
- Purity:Greater than 95% as determined by SDS-PAGE.
- Formulation:PKC-a is supplied at of 0.3mg/ml in 50mM Tris, pH 7.4, 0.1M NaCl, 20% glycerol, 1mM DTT, 0.1mM EDTA, 0.2 um PMSF and 0.03% Brij-35.
- Tested applications:SDS-PAGE
Specifications
Frequently Bought Together